missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4'-fluoroacetophenone, 99%, ACROS Organics™

Manufacturer:  Acros Organics 158170250

 View more versions of this product

Catalog No. AC158170250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-4'-fluoroacetophenone
Glass bottle
47°C to 50°C
456-04-2
100.0
C8H6ClFO
MFCD00011652
UJZWJOQRSMOFMA-UHFFFAOYSA-N
2-chloro-1-(4-fluorophenyl)ethanone
120248
Flakes or Platelets
Authentic
Undesignated
25g
98.5
99%
FC6H4COCH2Cl
2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone
C1=CC(=CC=C1C(=O)CCl)F
172.583
172.59
≥98.5%

Chemical Identifiers

456-04-2
172.583
UJZWJOQRSMOFMA-UHFFFAOYSA-N
120248
C1=CC(=CC=C1C(=O)CCl)F
C8H6ClFO
MFCD00011652
2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone
2-chloro-1-(4-fluorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Wear respiratory protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents