Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloro-4'-fluoroacetophenone, 98+%

$79.12 - $260.14

Chemical Identifiers

Chemical Name or Material 2'-Chloro-4'-fluoroacetophenone
CAS 700-35-6
Molecular Formula C8H6ClFO
Formula Weight 172.59
MDL Number MFCD00042202
Beilstein 2249796
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1988003

  SDS
alfa aesar™
L1988003
1g Each for $79.12

AAL1988006

  SDS
alfa aesar™
L1988006
5g Each for $260.14
Description & Specifications

Specifications

Formula Weight 172.59
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2'-Chloro-4'-fluoroacetophenone
Assay ≥98%
Molecular Formula C8H6ClFO
MDL Number MFCD00042202
CAS 700-35-6
Beilstein 2249796
Boiling Point 110°C (19mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.