Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4'-fluoroacetanilide, 98%

$55.55 - $198.57

Chemical Identifiers

Chemical Name or Material 2-Chloro-4'-fluoroacetanilide
CAS 351-04-2
Molecular Formula C8H7ClFNO
Formula Weight 187.6
MDL Number MFCD00015730
Synonym N-Chloroacetyl-4-fluoroaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1766906

  SDS
alfa aesar™
A1766906
5g Each for $55.55

AAA1766914

  SDS
alfa aesar™
A1766914
25g Each for $198.57
Description & Specifications

Specifications

Formula Weight 187.6
Chemical Name or Material 2-Chloro-4'-fluoroacetanilide
Assay 98%
Molecular Formula C8H7ClFNO
MDL Number MFCD00015730
CAS 351-04-2
Synonym N-Chloroacetyl-4-fluoroaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.