Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloro-4'-fluoroacetanilide, 98%

$137.12 - $433.95

Chemical Identifiers

Chemical Name or Material 2'-Chloro-4'-fluoroacetanilide
CAS 399-35-9
Molecular Formula C8H7ClFNO
Formula Weight 187.6
MDL Number MFCD00042594
Beilstein 2719131
Synonym N-(2-Chloro-4-fluorophenyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2176214

  SDS
alfa aesar™
B2176214
25g Each for $137.12

AAB2176222

  SDS
alfa aesar™
B2176222
100g Each for $433.95
Description & Specifications

Specifications

Formula Weight 187.6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Melting Point 116° to 118°C
Chemical Name or Material 2'-Chloro-4'-fluoroacetanilide
CAS 399-35-9
MDL Number MFCD00042594
Synonym N-(2-Chloro-4-fluorophenyl)acetamide
Beilstein 2719131
Assay 98%
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H7ClFNO
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.