missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-fluoro-1-iodobenzene, 98%

Manufacturer:  Alfa Aesar™ A1201006

 View more versions of this product

Catalog No. AAA1201006


 

Specifications

Specifications

2-Chloro-4-fluoro-1-iodobenzene
5g
98%
MFCD00051774
POTCKVPDYXEGSV-UHFFFAOYSA-N
2-chloro-4-fluoro-1-iodobenzene
2736559
96°C to 98°C (15mmHg)
101335-11-9
C6H3ClFI
2435080
C1=CC(=C(C=C1F)Cl)I
256.443
256.45

Chemical Identifiers

101335-11-9
256.443
POTCKVPDYXEGSV-UHFFFAOYSA-N
C6H3ClFI
MFCD00051774
2736559
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents