Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-cyanobenzenesulfonyl chloride, 95%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-cyanobenzenesulfonyl chloride
CAS 254749-11-6
Molecular Formula C7H3Cl2NO2S
Formula Weight 236.08
MDL Number MFCD00177089
UN Number UN2923
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5037903

  SDS
alfa aesar™
H5037903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50379MD

  SDS
alfa aesar™
H50379MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.08
Chemical Name or Material 2-Chloro-4-cyanobenzenesulfonyl chloride
Assay 95%
Molecular Formula C7H3Cl2NO2S
MDL Number MFCD00177089
CAS 254749-11-6
UN Number UN2923
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Melting Point 101° to 105°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.