Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4,8-dimethylquinoline, 98%

$313.41 - $313.41

Chemical Identifiers

Chemical Name or Material 2-Chloro-4,8-dimethylquinoline
CAS 3913-17-5
Molecular Formula C11H10ClN
Formula Weight 191.66
MDL Number MFCD00760267
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5015103

  SDS
alfa aesar™
H5015103
1g Each for $313.41
Description & Specifications

Specifications

Formula Weight 191.66
Molecular Formula C11H10ClN
MDL Number MFCD00760267
Quantity 1g
Assay 98%
CAS 3913-17-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Chloro-4,8-dimethylquinoline

Applications
2-Chloro-4,8-dimethylquinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.