Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4,6-dimethylpyrimidine, 99%

$332.66 - $332.66

Chemical Identifiers

Chemical Name or Material 2-Chloro-4,6-dimethylpyrimidine
CAS 4472-44-0
Molecular Formula C6H7ClN2
Formula Weight 142.59g/mol
MDL Number MFCD00023199
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5033103

 
alfa aesar™
H50331-03
Each for $332.66
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 2-Chloro-4,6-dimethylpyrimidine
CAS 4472-44-0
Assay 99%
Molecular Formula C6H7ClN2
Formula Weight 142.59g/mol
MDL Number MFCD00023199
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UV8000000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.