missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-4,6-dimethylpyrimidine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442550050
Specifications
2-Chloro-4, 6-dimethylpyrimidine | |
210°C to 212°C | |
36°C to 38°C | |
4472-44-0 | |
100.0 | |
C6H7ClN2 | |
RZVPFDOTMFYQHR-UHFFFAOYSA-N | |
2-chloro-4,6-dimethylpyrimidine | |
20550 | |
Powder, Crystalline Powder or Low Melting Solid |
Authentic | |
White to Yellow | |
5g | |
96.0 | |
96% min. (GC) | |
2-chlor-4,6-dimethylpyrimidine, 2-chloro-4,6-dimethyl-1,3-diazine, 2-chloro-4,6-dimethyl-pyrimidine, 2-chloro-4,6-dimethylpyrimidine, 4,6-dimethyl-2-chloropyrimidine, 5-23-05-00401 beilstein handbook reference, acmc-1adw7, ksc495i5r, pubchem6904, pyrimidine, 2-chloro-4,6-dimethyl | |
CC1=CC(=NC(=N1)Cl)C | |
142.59 | |
142.59 | |
97% |
Chemical Identifiers
4472-44-0 | |
142.59 | |
2-chlor-4,6-dimethylpyrimidine, 2-chloro-4,6-dimethyl-1,3-diazine, 2-chloro-4,6-dimethyl-pyrimidine, 2-chloro-4,6-dimethylpyrimidine, 4,6-dimethyl-2-chloropyrimidine, 5-23-05-00401 beilstein handbook reference, acmc-1adw7, ksc495i5r, pubchem6904, pyrimidine, 2-chloro-4,6-dimethyl | |
2-chloro-4,6-dimethylpyrimidine |
C6H7ClN2 | |
RZVPFDOTMFYQHR-UHFFFAOYSA-N | |
20550 | |
CC1=CC(=NC(=N1)Cl)C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning