Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4,6-bis(trifluoromethyl)benzaldehyde, 97%

$111.91 - $111.91

Chemical Identifiers

Chemical Name or Material 2-Chloro-4,6-bis(trifluoromethyl)benzaldehyde
Molecular Formula C9H3ClF6O
Formula Weight 276.56
MDL Number MFCD09025366
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH31532MD

  SDS
alfa aesar™
H31532MD
250mg Each for $111.91
Description & Specifications

Specifications

Formula Weight 276.56
Chemical Name or Material 2-Chloro-4,6-bis(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C9H3ClF6O
MDL Number MFCD09025366
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.