Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine, 96%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine
CAS 122-34-9
Molecular Formula C7H12ClN5
Formula Weight 201.66
MDL Number MFCD00023174
UN Number UN3077
Beilstein 10895
Merck Index 14,8533
Synonym Simazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0394722

  SDS
alfa aesar™
L0394722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.66
CAS 122-34-9
Chemical Name or Material 2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3077
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Molecular Formula C7H12ClN5
Quantity 100g
MDL Number MFCD00023174
Melting Point 227° to 229°C
TSCA TSCA
Synonym Simazine
RTECS Number XY5250000
Beilstein 10895
Merck Index 14,8533
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Assay 96%
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.