Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4,5-dimethylphenol, 98%


Quantity 1g

Manufacturer: alfa aesar™  L0162703

 View more versions of this product

Catalog No. AAL0162703

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 156.61
Chemical Name or Material 2-Chloro-4,5-dimethylphenol
Assay 98%
Molecular Formula C8H9ClO
MDL Number MFCD00002173
CAS 1124-04-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2042057
Synonym 6-Chloro-3,4-xylenol
Melting Point 70° to 72°C
Quantity 1g

Specifications

Chemical Name or Material 2-Chloro-4,5-dimethylphenol
CAS 1124-04-5
Molecular Formula C8H9ClO
Formula Weight 156.61
MDL Number MFCD00002173
Beilstein 2042057
Synonym 6-Chloro-3,4-xylenol
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.