Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4,5-difluorophenylacetic acid, 97%

$89.80 - $221.50

Chemical Identifiers

Chemical Name or Material 2-Chloro-4,5-difluorophenylacetic acid
CAS 883502-00-9
Molecular Formula C8H5ClF2O2
Formula Weight 206.58
MDL Number MFCD04973421
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2656603

  SDS
alfa aesar™
H2656603
1g Each for $221.50

AAH26566MD

  SDS
alfa aesar™
H26566MD
250mg Each for $89.80
Description & Specifications

Specifications

Formula Weight 206.58
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 99° to 101°C
Molecular Formula C8H5ClF2O2
Assay 97%
CAS 883502-00-9
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
MDL Number MFCD04973421
Health Hazard 1 Warning
Chemical Name or Material 2-Chloro-4,5-difluorophenylacetic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.