Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(4-morpholinyl)quinazoline, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-(4-morpholinyl)quinazoline
CAS 39213-05-3
Formula Weight 249.7
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6312903

 
alfa aesar™
H6312903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63129MD

 
alfa aesar™
H63129MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.7
Chemical Name or Material 2-Chloro-4-(4-morpholinyl)quinazoline
Quantity 1g
Physical Form Solid
Assay 97%
CAS 39213-05-3