Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(3-piperidinyloxy)benzonitrile, 97%

$389.43 - $389.43

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-(3-piperidinyloxy)benzonitrile
CAS 902836-92-4
Molecular Formula C12H13ClN2O
Formula Weight 236.7
MDL Number MFCD08061078
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5093803

 
alfa aesar™
H5093803
1g Each for $389.43
Description & Specifications

Specifications

Formula Weight 236.7
Chemical Name or Material 2-Chloro-4-(3-piperidinyloxy)benzonitrile
Assay 97%
Molecular Formula C12H13ClN2O
MDL Number MFCD08061078
CAS 902836-92-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.