missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5531603

 View more versions of this product

Catalog No. AAH5531603



Specifications

Specifications

2-Chloro-4-(1-pyrrolidinylcarbonyl)benzeneboronic acid
1449131-93-4
MFCD22125137
CCPHBMWIZPSWRQ-UHFFFAOYSA-N
[2-chloro-4-(pyrrolidine-1-carbonyl)phenyl]boronic acid
73996117
97%
1g
C11H13BClNO3
2-chloro-4-1-pyrrolidinylcarbonyl benzeneboronic acid
B(C1=C(C=C(C=C1)C(=O)N2CCCC2)Cl)(O)O
253.489
253.49

Chemical Identifiers

1449131-93-4
253.489
CCPHBMWIZPSWRQ-UHFFFAOYSA-N
73996117
B(C1=C(C=C(C=C1)C(=O)N2CCCC2)Cl)(O)O
C11H13BClNO3
MFCD22125137
2-chloro-4-1-pyrrolidinylcarbonyl benzeneboronic acid
[2-chloro-4-(pyrrolidine-1-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents