Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-(trifluoromethyl)pyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-(trifluoromethyl)pyridine
CAS 65753-47-1
Molecular Formula C6H3ClF3N
Formula Weight 181.55
MDL Number MFCD00042223
UN Number UN2923
Beilstein 4179716
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0830303

 
alfa aesar™
L0830303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0830306

 
alfa aesar™
L0830306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.55
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)pyridine
Assay 98%
Molecular Formula C6H3ClF3N
MDL Number MFCD00042223
CAS 65753-47-1
UN Number UN2923
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H310-H372-H314-H318
Toxic if swallowed.
Fatal in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P261-P280-P273-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Melting Point 37° to 40°C
Beilstein 4179716
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Boiling Point 166° to 168°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P273-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H310-H372-H314-H318
Toxic if swallowed.
Fatal in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.