missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 241790250

 View more versions of this product

Catalog No. AC241790250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-chloro-3-(trifluoromethyl)pyridine
96.0
166.0°C to 168.0°C
Authentic
Glass bottle
25g
Solubility in water: insoluble. Other solubilities: reacts at high temperature with strong acids and,alkalis
C1=CC(=C(N=C1)Cl)C(F)(F)F
181.54
181.54
65753-47-1
100.0
82°C
96% min. (GC)
C6H3ClF3N
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t
RXATZPCCMYMPME-UHFFFAOYSA-N
2-chloro-3-(trifluoromethyl)pyridine
589833
97%

Chemical Identifiers

65753-47-1
181.54
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t
2-chloro-3-(trifluoromethyl)pyridine
C6H3ClF3N
RXATZPCCMYMPME-UHFFFAOYSA-N
589833
C1=CC(=C(N=C1)Cl)C(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Harmful to aquatic life with long lasting e

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only