Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-(trifluoromethyl)phenylacetic acid, 97%

$183.82 - $183.82

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-(trifluoromethyl)phenylacetic acid
CAS 22902-84-7
Molecular Formula C9H6ClF3O2
Formula Weight 238.59
MDL Number MFCD04115992
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2677703

  SDS
alfa aesar™
H2677703
1g Each for $183.82

AAH26777MD

  SDS
alfa aesar™
H26777MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Chloro-3-(trifluoromethyl)phenylacetic acid
Melting Point 131° to 133°C
Quantity 1g
CAS 22902-84-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Assay 97%
Molecular Formula C9H6ClF3O2
Formula Weight 238.59
MDL Number MFCD04115992
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.