Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-(trifluoromethyl)phenol, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-(trifluoromethyl)phenol
CAS 138377-34-1
Molecular Formula C7H4ClF3O
Formula Weight 196.56
MDL Number MFCD00213793
UN Number UN3265
Beilstein 9045773
Synonym 2-Chloro-3-hydroxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1932303

  SDS
alfa aesar™
L1932303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1932306

  SDS
alfa aesar™
L1932306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.56
CAS 138377-34-1
Assay 98%
Molecular Formula C7H4ClF3O
MDL Number MFCD00213793
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Synonym 2-Chloro-3-hydroxybenzotrifluoride
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN3265
Quantity 1g
Health Hazard 1 Danger
Flash Point 79°C (174°F)
Boiling Point 160°C
Beilstein 9045773
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)phenol
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.