Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-(trifluoromethyl)benzyl alcohol, 97+%

$84.81 - $303.24

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzyl alcohol
CAS 261763-20-6
Molecular Formula C8H6ClF3O
Formula Weight 210.58
MDL Number MFCD01631537
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2321203

 
alfa aesar™
B2321203
1g Each for $84.81

AAB2321206

 
alfa aesar™
B2321206
5g Each for $303.24
Description & Specifications

Specifications

Formula Weight 210.58
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzyl alcohol
Assay ≥97%
Molecular Formula C8H6ClF3O
MDL Number MFCD01631537
CAS 261763-20-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 74° to 77°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.