Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-(trifluoromethyl)benzoic acid, 97+%

$272.73 - $272.73

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzoic acid
CAS 39226-97-6
Molecular Formula C8H4ClF3O2
Formula Weight 224.57
MDL Number MFCD00792415
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2274406

  SDS
alfa aesar™
B2274406
5g Each for $272.73
Description & Specifications

Specifications

Formula Weight 224.57
MDL Number MFCD00792415
Molecular Formula C8H4ClF3O2
Assay ≥97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
CAS 39226-97-6
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 132° to 136°C
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.