2-Chloro-3-(trifluoromethyl)benzoic acid, 97+%
Manufacturer:  Alfa Aesar B2274403
 View more versions of this product
Catalog No. AAB2274403
Specifications
Specifications
2-Chloro-3-(trifluoromethyl)benzoic acid
1g
≥97%
MFCD00792415
C1=CC(=C(C(=C1)C(F)(F)F)Cl)C(=O)O
224.563
224.57
132° to 136°C
39226-97-6
C8H4ClF3O2
AXOAWWUSRZCGKS-UHFFFAOYSA-N
2-chloro-3-(trifluoromethyl)benzoic acid
2736677
Chemical Identifiers
39226-97-6
224.563
AXOAWWUSRZCGKS-UHFFFAOYSA-N
C8H4ClF3O2
MFCD00792415
2736677
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
SDS Documents