Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-(trifluoromethyl)aniline
CAS 62476-58-8
Molecular Formula C7H5ClF3N
Formula Weight 195.57
MDL Number MFCD04115988
UN Number UN2811
Synonym 3-Amino-2-chlorobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2678103

 
alfa aesar™
H2678103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2678106

 
alfa aesar™
H2678106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.57
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)aniline
Assay 97%
Molecular Formula C7H5ClF3N
MDL Number MFCD04115988
CAS 62476-58-8
UN Number UN2811
Synonym 3-Amino-2-chlorobenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 25° to 28°C
Quantity 1g

Applications
As pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.