missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-3-pyridinemethanol 98.0+%, TCI America™
$44.79 - $134.38
Chemical Identifiers
CAS | 42330-59-6 |
---|---|
Molecular Formula | C6H6ClNO |
Molecular Weight (g/mol) | 143.57 |
MDL Number | MFCD01935961 |
InChI Key | HMPDWSBKPCOQDW-UHFFFAOYSA-N |
Synonym | 2-chloropyridin-3-yl methanol, 2-chloro-3-pyridinyl methanol, 2-chloro-3-pyridinylmethanol, 2-chloro-3-pyridinemethanol, 2-chloro-3-hydroxymethyl pyridine, 2-chloro-pyridin-3-yl-methanol, 3-pyridinemethanol, 2-chloro, 2-chloro-3-hydroxymethylpyridine |
PubChem CID | 2763647 |
IUPAC Name | (2-chloropyridin-3-yl)methanol |
SMILES | OCC1=CC=CN=C1Cl |
Chemical Identifiers
42330-59-6 | |
143.57 | |
HMPDWSBKPCOQDW-UHFFFAOYSA-N | |
2763647 | |
OCC1=CC=CN=C1Cl |
C6H6ClNO | |
MFCD01935961 | |
2-chloropyridin-3-yl methanol, 2-chloro-3-pyridinyl methanol, 2-chloro-3-pyridinylmethanol, 2-chloro-3-pyridinemethanol, 2-chloro-3-hydroxymethyl pyridine, 2-chloro-pyridin-3-yl-methanol, 3-pyridinemethanol, 2-chloro, 2-chloro-3-hydroxymethylpyridine | |
(2-chloropyridin-3-yl)methanol |
Specifications
42330-59-6 | |
112°C | |
MFCD01935961 | |
2-chloropyridin-3-yl methanol, 2-chloro-3-pyridinyl methanol, 2-chloro-3-pyridinylmethanol, 2-chloro-3-pyridinemethanol, 2-chloro-3-hydroxymethyl pyridine, 2-chloro-pyridin-3-yl-methanol, 3-pyridinemethanol, 2-chloro, 2-chloro-3-hydroxymethylpyridine | |
OCC1=CC=CN=C1Cl | |
143.57 | |
143.57 | |
Crystalline Lumps |
63°C | |
C6H6ClNO | |
1 g | |
HMPDWSBKPCOQDW-UHFFFAOYSA-N | |
(2-chloropyridin-3-yl)methanol | |
2763647 | |
≥98.0% (GC) | |
2-Chloro-3-pyridinemethanol |
Safety and Handling
TSCA : No