Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-pyridinemethanol, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-pyridinemethanol
CAS 42330-59-6
Formula Weight 143.57
MDL Number MFCD01935961
Beilstein 1525378
Synonym 2-Chloro-3-(hydroxymethyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6334103

  SDS
alfa aesar™
H6334103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6334106

  SDS
alfa aesar™
H6334106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 61° to 65°C (dec.)
Formula Weight 143.57
CAS 42330-59-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Chloro-3-pyridinemethanol
Physical Form Solid
Assay 97%
Synonym 2-Chloro-3-(hydroxymethyl)pyridine
MDL Number MFCD01935961
Storage Note 1 2° to 8°C
Beilstein 1525378
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Boiling Point 112°C (0.2mmHg)
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C