missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-chloro-3-picoline, 98%, Thermo Scientific™
$124.87 - $124.87
Chemical Identifiers
CAS | 18368-76-8 |
---|---|
Molecular Formula | C6H6ClN |
Molecular Weight (g/mol) | 127.57 |
MDL Number | MFCD00234177 |
InChI Key | RKVUCIFREKHYTL-UHFFFAOYSA-N |
PubChem CID | 87603 |
IUPAC Name | 2-chloro-3-methylpyridine |
SMILES | CC1=C(N=CC=C1)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC379580050
|
Thermo Scientific Chemicals
379580050 |
5 g | Glass bottle |
Each for $124.87
|
|
||||
Chemical Identifiers
18368-76-8 | |
127.57 | |
RKVUCIFREKHYTL-UHFFFAOYSA-N | |
2-chloro-3-methylpyridine |
C6H6ClN | |
MFCD00234177 | |
87603 | |
CC1=C(N=CC=C1)Cl |
Specifications
18368-76-8 | |
100.0 | |
1.1700g/mL | |
79°C | |
97.5% min. (GC) | |
C6H6ClN | |
MFCD00234177 | |
1.17 | |
RKVUCIFREKHYTL-UHFFFAOYSA-N | |
2-chloro-3-methylpyridine | |
87603 | |
98% | |
2-Chloro-3-picoline |
97.5 | |
Colorless to Yellow | |
192°C to 193°C | |
Authentic | |
Glass bottle | |
1.5320 to 1.534 | |
5 g | |
2-chloro-3-picoline, 2-chloro 3-methylpyridine, 3-picoline, 2-chloro, pyridine, 2-chloro-3-methyl, 2-chloro-3-methyl-pyridine, pyridine,2-chloro-3-methyl, pubchem2074, 2-chloro-methyl-pyridine, 2-chloro3-methyl-pyridine | |
CC1=C(N=CC=C1)Cl | |
127.57 | |
127.57 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 242-242-1
Recommended Storage : this product may darken during storage
RUO – Research Use Only