missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-3-picoline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC379580250

 View more versions of this product

Catalog No. AC379580250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Chloro-3-picoline
Colorless to light yellow
18368-76-8
100.0
Glass bottle
this product may darken during storage
98%
C6H6ClN
MFCD00234177
1.1700g/mL
25g
97.5
Authentic
1.5320 to 1.5340 (20°C, 589nm)
Liquid
98%
127.57
2-Chloro-3-methylpyridine

Chemical Identifiers

18368-76-8
100.0
C6H6ClN
MFCD00234177
2-Chloro-3-picoline
97.5
98%
127.57
2-Chloro-3-methylpyridine
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning