missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-nitropyridine, 99%

Manufacturer:  Alfa Aesar™ A1216122

 View more versions of this product

Catalog No. AAA1216122


 

Specifications

Specifications

2-Chloro-3-nitropyridine
99°C to 103°C
5470-18-8
C5H3ClN2O2
124482
C1=CC(=C(N=C1)Cl)[N+](=O)[O-]
158.541
158.54
185°C (365°F)
100g
99%
MFCD00006232
UUOLETYDNTVQDY-UHFFFAOYSA-N
2-chloro-3-nitropyridine
79613

Chemical Identifiers

5470-18-8
158.541
UUOLETYDNTVQDY-UHFFFAOYSA-N
C5H3ClN2O2
MFCD00006232
79613
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

TSCA : TSCA

SDS missing translation for 'documents'