missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-nitrobenzotrifluoride, 98%

Manufacturer:  Alfa Aesar™ H6405114

 View more versions of this product

Catalog No. AAH6405114



Specifications

Specifications

2-Chloro-3-nitrobenzotrifluoride
25°C to 30°C
39974-35-1
MFCD14525509
2-chloro-1-nitro-3-trifluoromethyl benzene, 2-chloro-3-nitrobenzotrifluoride, 2-chloro-3-trifluoromethyl-1-nitrobenzene, 2-nitro-6-trifluoromethyl chlorobenzene, benzene, 2-chloro-1-nitro-3-trifluoromethyl, benzene,2-chloro-1-nitro-3-trifluoromethyl, o-chloronitrobenzotrifluoride
C1=CC(=C(C(=C1)[N+](=O)[O-])Cl)C(F)(F)F
225.551
225.55
98%
>110°C (230°F)
25g
C7H3ClF3NO2
10429463
TWABRHQBRWLSSE-UHFFFAOYSA-N
2-chloro-1-nitro-3-(trifluoromethyl)benzene
3016126
Solid

Chemical Identifiers

39974-35-1
225.551
TWABRHQBRWLSSE-UHFFFAOYSA-N
3016126
C1=CC(=C(C(=C1)[N+](=O)[O-])Cl)C(F)(F)F
C7H3ClF3NO2
MFCD14525509
2-chloro-1-nitro-3-trifluoromethyl benzene, 2-chloro-3-nitrobenzotrifluoride, 2-chloro-3-trifluoromethyl-1-nitrobenzene, 2-nitro-6-trifluoromethyl chlorobenzene, benzene, 2-chloro-1-nitro-3-trifluoromethyl, benzene,2-chloro-1-nitro-3-trifluoromethyl, o-chloronitrobenzotrifluoride
2-chloro-1-nitro-3-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 254-724-9

TSCA : No

Recommended Storage : Ambient temperatures

Documents