Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3'-nitroacetanilide, 98%

$49.11 - $601.20

Chemical Identifiers

Chemical Name or Material 2-Chloro-3'-nitroacetanilide
CAS 10147-71-4
Molecular Formula C8H7ClN2O3
Formula Weight 214.61
MDL Number MFCD00024359
Beilstein 913533
Synonym alpha-Chloro-3-nitroacetanilide; N-Chloroacetyl-3-nitroaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1262106

  SDS
alfa aesar™
A1262106
5g Each for $49.11

AAA1262122

  SDS
alfa aesar™
A1262122
100g Each for $601.20
Description & Specifications

Specifications

Formula Weight 214.61
MDL Number MFCD00024359
Health Hazard 1 Warning
CAS 10147-71-4
Molecular Formula C8H7ClN2O3
Melting Point 112° to 115°C
Synonym alpha-Chloro-3-nitroacetanilide; N-Chloroacetyl-3-nitroaniline
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Beilstein 913533
Assay 98%
Chemical Name or Material 2-Chloro-3'-nitroacetanilide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.