missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, 95%

Manufacturer:  Alfa Aesar™ H6439406

 View more versions of this product

Catalog No. AAH6439406



Specifications

Specifications

Colorless to Yellow
72587-15-6
MFCD06255171
2-chloro-3-nitro-5-trifluoromethyl pyridine, 2-chloro-3-nitro-5-trifluoromethyl-pyridine, 2-chloro-5-trifluoromethyl-3-nitropyridine, 2-chloro3-nitro-5-trifluoromethyl pyridine, 5-trifluoromethyl-3-nitro-2-chloropyridine, abbypharma ap-11-5568, acmc-209onn, pubchem17529, pyridine, 2-chloro-3-nitro-5-trifluoromethyl
C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F
226.539
226.54
95%
5g
C6H2ClF3N2O2
7586778
DRPIKFKCAJGTJF-UHFFFAOYSA-N
2-chloro-3-nitro-5-(trifluoromethyl)pyridine
4736950
Liquid

Chemical Identifiers

72587-15-6
226.539
DRPIKFKCAJGTJF-UHFFFAOYSA-N
4736950
C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F
C6H2ClF3N2O2
MFCD06255171
2-chloro-3-nitro-5-trifluoromethyl pyridine, 2-chloro-3-nitro-5-trifluoromethyl-pyridine, 2-chloro-5-trifluoromethyl-3-nitropyridine, 2-chloro3-nitro-5-trifluoromethyl pyridine, 5-trifluoromethyl-3-nitro-2-chloropyridine, abbypharma ap-11-5568, acmc-209onn, pubchem17529, pyridine, 2-chloro-3-nitro-5-trifluoromethyl
2-chloro-3-nitro-5-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents