Please login to your online account to display your discounted pricing

2-Chloro-3-nitro-5-picoline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-nitro-5-picoline
CAS 23056-40-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H5 Cl N2 O2
Formula Weight 172.57
MDL Number MFCD02070020
Synonym 2-Chloro-5-methyl-3-nitropyridine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379540010

  SDS
thermo fisher scientific
AC379540010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 172.57
Quantity 1g
Color Beige
Melting Point 46° to 50°C
CAS Max % 100.0
Synonym 2-Chloro-5-methyl-3-nitropyridine
Infrared Spectrum Authentic
Chemical Name or Material 2-Chloro-3-nitro-5-picoline
MDL Number MFCD02070020
CAS 23056-40-8
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 97.5
Molecular Formula C6 H5 Cl N2 O2
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%