missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-methylthiophene, 97%

$45.32 - $45.32

Chemical Identifiers

CAS 14345-97-2
Molecular Formula C5H5ClS
Formula Weight 132.61
MDL Number MFCD00130083
UN Number UN1993
Chemical Name or Material 2-Chloro-3-methylthiophene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2795703 SDS Alfa Aesar™
H2795703
1g Each for $45.32
AAH2795706 SDS Alfa Aesar™
H2795706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH2795714 SDS Alfa Aesar™
H2795714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

14345-97-2
132.61
UN1993
C5H5ClS
MFCD00130083
2-Chloro-3-methylthiophene

Specifications

46°C (115°F)
1g
C5H5ClS
MFCD00130083
2-Chloro-3-methylthiophene
147° to 149°C
14345-97-2
132.61
UN1993
97%

Safety and Handling

GHS H Statement
H301-H311-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.