Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-methylquinoxaline, 95%

$108.38 - $312.47

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-methylquinoxaline
CAS 32601-86-8
Molecular Formula C9H7ClN2
Formula Weight 178.62
MDL Number MFCD00023913
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5173403

 
alfa aesar™
H5173403
1g Each for $312.47

AAH51734MD

 
alfa aesar™
H51734MD
250mg Each for $108.38
Description & Specifications

Specifications

Formula Weight 178.62
Chemical Name or Material 2-Chloro-3-methylquinoxaline
Assay 95%
Molecular Formula C9H7ClN2
MDL Number MFCD00023913
CAS 32601-86-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 71° to 75°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.