Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-methylquinoline, 97%

$135.61 - $135.61

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-methylquinoline
CAS 57876-69-4
Molecular Formula C10H8ClN
Formula Weight 177.63
MDL Number MFCD00100265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5049203

  SDS
alfa aesar™
H5049203
1g Each for $135.61

AAH5049206

  SDS
alfa aesar™
H5049206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Chloro-3-methylquinoline
Melting Point 82° to 84°C
Quantity 1g
CAS 57876-69-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
Molecular Formula C10H8ClN
Formula Weight 177.63
MDL Number MFCD00100265

Applications
2-Chloro-3-methylquinoline is used as organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.