Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-methylpyridine, 98+%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-methylpyridine
CAS 18368-76-8
Molecular Formula C6H6ClN
Formula Weight 127.57
MDL Number MFCD00234177
UN Number UN2810
Beilstein 107895
Synonym 2-Chloro-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1986114

  SDS
alfa aesar™
L1986114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1986106

  SDS
alfa aesar™
L1986106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.57
CAS 18368-76-8
MDL Number MFCD00234177
Chemical Name or Material 2-Chloro-3-methylpyridine
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2810
Molecular Formula C6H6ClN
Beilstein 107895
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 25g
Boiling Point 192° to 193°C
Flash Point 79°C (174°F)
Synonym 2-Chloro-3-picoline
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.