Please login to your online account to display your discounted pricing

2-Chloro-3-methylbenzotrifluoride, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-methylbenzotrifluoride
Assay Percent Range 95%
Molecular Formula C8H6ClF3
Formula Weight 194.58
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC451290010

  SDS
thermo fisher scientific
AC451290010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC451290050

  SDS
thermo fisher scientific
AC451290050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.58
Quantity 1g
Chemical Name or Material 2-Chloro-3-methylbenzotrifluoride
Assay Percent Range 95%
Molecular Formula C8H6ClF3
CAS 74483-48-0