missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-3-methyl-5-nitropyridine, 97%, Maybridge

$15.71 - $15.71

Chemical Identifiers

CAS 22280-56-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H5ClN2O2
Formula Weight 172.57
MDL Number MFCD00173686
Synonym 2-Chloro-3-methyl-5-nitropyridine
Chemical Name or Material 2-Chloro-3-methyl-5-nitropyridine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BTB09487DA SDS Fisher Scientific Company
BTB09487DA
Amber Glass Bottle 1g Each for $15.71
Specifications Description & Specifications

Chemical Identifiers

22280-56-4
100.0
C6H5ClN2O2
MFCD00173686
2-Chloro-3-methyl-5-nitropyridine
96.5
97%
172.57
2-Chloro-3-methyl-5-nitropyridine
97%

Specifications

22280-56-4
100.0
97%
C6H5ClN2O2
MFCD00173686
2-Chloro-3-methyl-5-nitropyridine
96.5
Amber Glass Bottle
1g
172.57
2-Chloro-3-methyl-5-nitropyridine
97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning