missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-methoxypyridine, 97%

$117.05 - $430.26

Chemical Identifiers

CAS 52605-96-6
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD03426022
InChI Key CCVNZKGBNUPYPG-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3-methoxy, 2-chloro-3-methoxy-pyridine, pubchem6146, acmc-1ap1j, 2chloro-3-methoxylpyridine, ksc498a8t
PubChem CID 104252
IUPAC Name 2-chloro-3-methoxypyridine
SMILES COC1=C(N=CC=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1769603 View Documents Alfa Aesar™
L1769603
1g Each for $117.05
 
AAL1769606 View Documents Alfa Aesar™
L1769606
5g Each for $430.26
 
Description

Description

Applications
It is a pharmaceutical raw material and also used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

52605-96-6
143.57
CCVNZKGBNUPYPG-UHFFFAOYSA-N
104252
COC1=C(N=CC=C1)Cl
C6H6ClNO
MFCD03426022
pyridine, 2-chloro-3-methoxy, 2-chloro-3-methoxy-pyridine, pubchem6146, acmc-1ap1j, 2chloro-3-methoxylpyridine, ksc498a8t
2-chloro-3-methoxypyridine

Specifications

46°C to 48°C
52605-96-6
MFCD03426022
pyridine, 2-chloro-3-methoxy, 2-chloro-3-methoxy-pyridine, pubchem6146, acmc-1ap1j, 2chloro-3-methoxylpyridine, ksc498a8t
COC1=C(N=CC=C1)Cl
143.57
143.57
2-Chloro-3-methoxypyridine
1g
C6H6ClNO
115568
CCVNZKGBNUPYPG-UHFFFAOYSA-N
2-chloro-3-methoxypyridine
104252
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning