Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-iodopyridine, 97%

$96.43 - $96.43

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-iodopyridine
CAS 78607-36-0
Molecular Formula C5H3ClIN
Formula Weight 239.44
MDL Number MFCD00661298
Beilstein 4243304
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2002803

  SDS
alfa aesar™
L2002803
1g Each for $96.43
Description & Specifications

Specifications

Formula Weight 239.44
Chemical Name or Material 2-Chloro-3-iodopyridine
Assay 97%
Molecular Formula C5H3ClIN
MDL Number MFCD00661298
CAS 78607-36-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 4243304
Melting Point 93° to 97°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.