missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-hydroxypyridine, 98%

Manufacturer:  Alfa Aesar™ A1137606

 View more versions of this product

Catalog No. AAA1137606


 

Specifications

Specifications

2-Chloro-3-hydroxypyridine
5g
98%
MFCD00006235
2-Chloro-3-pyridinol
C1=CC(=C(N=C1)Cl)O
129.543
129.55
168° to 172°C
6636-78-8
C5H4ClNO
2216
RSOPTYAZDFSMTN-UHFFFAOYSA-N
2-chloropyridin-3-ol
81136

Chemical Identifiers

6636-78-8
129.543
RSOPTYAZDFSMTN-UHFFFAOYSA-N
81136
C5H4ClNO
MFCD00006235
2-Chloro-3-pyridinol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UU7707000

SDS Documents