missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-3-fluoropyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 17282-04-1
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.54
InChI Key SVAZIMBLBHOVIR-UHFFFAOYSA-N
Synonym 2-chloro-3-fluoro pyridine, 2-chloro-3-fluoro-pyridine, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g, 3-fluoro-2-chloropyridine, acmc-1cc0d, ksc174s3h, pubchem2532, pyridine, 2-chloro-3-fluoro
PubChem CID 2782791
IUPAC Name 2-chloro-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442080050
SDS View Documents
Thermo Fisher Scientific
AC442080050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442080250
View Documents
Thermo Fisher Scientific
AC442080250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

17282-04-1
131.54
2-chloro-3-fluoro pyridine, 2-chloro-3-fluoro-pyridine, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g, 3-fluoro-2-chloropyridine, acmc-1cc0d, ksc174s3h, pubchem2532, pyridine, 2-chloro-3-fluoro
2-chloro-3-fluoropyridine
C5H3ClFN
SVAZIMBLBHOVIR-UHFFFAOYSA-N
2782791
C1=CC(=C(N=C1)Cl)F

Specifications

1.3230g/mL
Glass bottle
80°C (80.0mmHg)
17282-04-1
C5H3ClFN
SVAZIMBLBHOVIR-UHFFFAOYSA-N
2-chloro-3-fluoropyridine
2782791
97%
63°C
1.323
5g
96% min. (GC)
2-chloro-3-fluoro pyridine, 2-chloro-3-fluoro-pyridine, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g, 3-fluoro-2-chloropyridine, acmc-1cc0d, ksc174s3h, pubchem2532, pyridine, 2-chloro-3-fluoro
C1=CC(=C(N=C1)Cl)F
131.54
131.54
2-Chloro-3-fluoropyridine
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS