Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-fluoropyridine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-fluoropyridine
CAS 17282-04-1
Molecular Formula C5H3ClFN
Formula Weight 131.54
MDL Number MFCD03095302
UN Number UN2810
Beilstein 1363435
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2002203

  SDS
alfa aesar™
L2002203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2002206

  SDS
alfa aesar™
L2002206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 131.54
Molecular Formula C5H3ClFN
Chemical Name or Material 2-Chloro-3-fluoropyridine
MDL Number MFCD03095302
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H301-H310-H330-H318-H227-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Odor Unpleasant
Quantity 1g
Flash Point 72°C (161°F)
CAS 17282-04-1
Assay 97%
Boiling Point 62° to 64°C (23mmHg)
UN Number UN2810
Health Hazard 1 Danger
Beilstein 1363435
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H310-H330-H318-H227-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.