Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-fluoro-6-methylpyridine, 95%

$156.70 - $156.70

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-fluoro-6-methylpyridine
CAS 374633-32-6
Molecular Formula C6H5ClFN
Formula Weight 145.56
MDL Number MFCD03095216
Beilstein 22770804
Synonym 2-Chloro-3-fluoro-6-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6447303

  SDS
alfa aesar™
H6447303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64473MD

  SDS
alfa aesar™
H64473MD
250mg Each for $156.70
Description & Specifications

Specifications

Formula Weight 145.56
Molecular Formula C6H5ClFN
Synonym 2-Chloro-3-fluoro-6-picoline
Chemical Name or Material 2-Chloro-3-fluoro-6-methylpyridine
MDL Number MFCD03095216
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 22770804
Quantity 1g
CAS 374633-32-6
Assay 95%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.