Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine
CAS 628692-22-8
Molecular Formula C6H2ClF4N
Formula Weight 199.54
MDL Number MFCD04038326
UN Number UN2810
Synonym 2-Chloro-alpha,alpha,alpha,3-tetrafluoro-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2002603

  SDS
alfa aesar™
L2002603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL20026MD

  SDS
alfa aesar™
L20026MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.54
Molecular Formula C6H2ClF4N
Chemical Name or Material 2-Chloro-3-fluoro-4-(trifluoromethyl)pyridine
CAS 628692-22-8
Assay 98%
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Quantity 1g
Boiling Point 35° to 37°C (25mmHg)
Synonym 2-Chloro-alpha,alpha,alpha,3-tetrafluoro-4-picoline
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD04038326
UN Number UN2810
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.