missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate, 97%, ACROS Organics™

Chemical Identifiers

CAS 63212-53-3
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H9ClNO·BF4
Formula Weight 269.42
MDL Number MFCD00011943
Chemical Name or Material 2-Chloro-3-ethylbenzoxazolium tetrafluoroborate, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC214000050 SDS Thermo Fisher Scientific
214000050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

63212-53-3
100.0
C9H9ClNO·BF4
MFCD00011943
≥96.0%
96.0
97%
269.42
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate, 97%

Specifications

Crystals or Crystalline Powder
5g
100.0
Authentic
C9H9ClNO·BF4
97%
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate, 97%
269.42
Beige
96.0
Glass bottle
63212-53-3
MFCD00011943
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 263-998-9

TSCA : TSCA