missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-cyanopyridine, 98%

Manufacturer:  Alfa Aesar™ A1481803

 View more versions of this product

Catalog No. AAA1481803



Specifications

Specifications

2-Chloro-3-cyanopyridine
105°C to 109°C
6602-54-6
MFCD00014628
115772
JAUPUQRPBNDMDT-UHFFFAOYSA-N
2-chloropyridine-3-carbonitrile
81079
98%
112°C (1mmHg)
1g
C6H3ClN2
UN3439
2-chloro-3-cyanopyridine, 2-chloronicotinonitrile, 2-chloro-3-pyridinecarbonitrile, 3-cyano-2-chloropyridine, 3-pyridinecarbonitrile, 2-chloro, 2-chloro-nicotinonitrile, 2-chloronicotinitrile, 2-chloro-3-cyano-pyridine, 2-chloro-3-cyanopyridine 2-chloronicotinonitrile, 2chloronicotinonitrile
C1=CC(=C(N=C1)Cl)C#N
138.554
138.56

Chemical Identifiers

6602-54-6
138.554
JAUPUQRPBNDMDT-UHFFFAOYSA-N
81079
C1=CC(=C(N=C1)Cl)C#N
C6H3ClN2
MFCD00014628
2-chloro-3-cyanopyridine, 2-chloronicotinonitrile, 2-chloro-3-pyridinecarbonitrile, 3-cyano-2-chloropyridine, 3-pyridinecarbonitrile, 2-chloro, 2-chloro-nicotinonitrile, 2-chloronicotinitrile, 2-chloro-3-cyano-pyridine, 2-chloro-3-cyanopyridine 2-chloronicotinonitrile, 2chloronicotinonitrile
2-chloropyridine-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents