missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester, 95%

$191.95 - $528.72

Chemical Identifiers

CAS 878194-94-6
Molecular Formula C12H14BClN2O2
Molecular Weight (g/mol) 264.516
MDL Number MFCD09037479
InChI Key YYPZHORJYDCOSQ-UHFFFAOYSA-N
Synonym 2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 44755174
IUPAC Name 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27284MD View Documents Alfa Aesar™
H27284MD
250mg Each for $191.95
 
AAH2728403 View Documents Alfa Aesar™
H2728403
1g Each for $528.72
 
Specifications

Chemical Identifiers

878194-94-6
264.516
YYPZHORJYDCOSQ-UHFFFAOYSA-N
44755174
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N
C12H14BClN2O2
MFCD09037479
2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile

Specifications

100°C to 101°C
878194-94-6
MFCD09037479
YYPZHORJYDCOSQ-UHFFFAOYSA-N
2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
44755174
95%
250mg
C12H14BClN2O2
2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N
264.516
264.52
2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures