missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-cyano-6-methylpyridine, 99%

Manufacturer:  Alfa Aesar™ A1564806

 View more versions of this product

Catalog No. AAA1564806



Specifications

Specifications

2-Chloro-3-cyano-6-methylpyridine
5g
C7H5ClN2
120414
YSBNBAYNISAUIT-UHFFFAOYSA-N
2-chloro-6-methylpyridine-3-carbonitrile
520400
99%
114°C to 117°C
28900-10-9
MFCD00082767
2-chloro-3-cyano-6-methyl pyridine
CC1=NC(=C(C=C1)C#N)Cl
152.581
152.58

Chemical Identifiers

28900-10-9
152.581
YSBNBAYNISAUIT-UHFFFAOYSA-N
520400
CC1=NC(=C(C=C1)C#N)Cl
C7H5ClN2
MFCD00082767
2-chloro-3-cyano-6-methyl pyridine
2-chloro-6-methylpyridine-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents